Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC American Formula All-Purpose Cleaner, Liquid, Hydrogen Peroxide, 1 gal

Catalog No. 502836077
Encompass

This liquid all-purpose cleaner has an orange scent. It is an environmentally responsible solution designed to quickly remove common soils and stains without harming the environment. By combining the versatile power of hydrogen peroxide with the cleaning ability of d-Limonene, it provides a safe and effective solution for all your day-to-day cleaning needs.

  • All-purpose cleaner
  • Liquid form
  • Bottle container type
  • 1 gal container size
  • Orange scent
  • Hydrogen peroxide formula type
  • Dilution: 1 oz per gallon or 10 oz per gallon
  • pH: 5.5-6.5
  • Colorless liquid
  • Origin: United States
  • Environmentally responsible
  • Quickly removes common soils and stains
  • Safe and effective solution

Catalog No. 50-283-6077 Supplier MSC Supplier No. 57209306
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This liquid all-purpose cleaner has an orange scent. It is an environmentally responsible solution designed to quickly remove common soils and stains without harming the environment. By combining the versatile power of hydrogen peroxide with the cleaning ability of d-Limonene, it provides a safe and effective solution for all your day-to-day cleaning needs.

  • All-purpose cleaner
  • Liquid form
  • Bottle container type
  • 1 gal container size
  • Orange scent
  • Hydrogen peroxide formula type
  • Dilution: 1 oz per gallon or 10 oz per gallon
  • pH: 5.5-6.5
  • Colorless liquid
  • Origin: United States
  • Environmentally responsible
  • Quickly removes common soils and stains
  • Safe and effective solution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.