Learn More
Oxyrase Oxyrase For Broth 100Ml Bottle 1/Pk
SDP
Catalog No. 502005300
Encompass
Oxyrase for Broth produces anaerobic conditions in wide variety of bacteriological broth media. Makes growing anaerobes easy; especially reviving lyophiles. Add Oxyrase for Broth to your broth medium. Broth goes anaerobic in 10-15 minutes with continual oxygen removal. Never boil broth tubes again.
This item is not returnable. View return policy
Catalog No. 50-200-5300 Supplier Oxyrase Supplier No. OB0100
Oxyrase for Broth produces anaerobic conditions in wide variety of bacteriological broth media. Makes growing anaerobes easy; especially reviving lyophiles. Add Oxyrase for Broth to your broth medium. Broth goes anaerobic in 10-15 minutes with continual oxygen removal. Never boil broth tubes again.