Learn More
Broadley James Corporation OXY II CARTRIDGE, 16PC
Catalog No. NC0339475
Encompass
16 Piece Cartridge Kit for OxyProbe IIThis convenient bulk pack is the most popular among large volume users. It contains 16 membrane cartridges in one easy-to-store box. Internal o-rings and electrolyte must be ordered separately.* OxyProbe� II replacement parts are not compatible with original OxyProbe� sensors
Catalog No. NC0339475 Supplier Broadley James Corporation Supplier No. MS2916
16 Piece Cartridge Kit for OxyProbe IIThis convenient bulk pack is the most popular among large volume users. It contains 16 membrane cartridges in one easy-to-store box. Internal o-rings and electrolyte must be ordered separately.* OxyProbe� II replacement parts are not compatible with original OxyProbe� sensors