Learn More
Broadley James Corporation D540-VP (12mm) OxyProbe II Dissolved Oxygen Sensor
Catalog No. NC0548926
Encompass
After several years of refinement we are introducing our latest set of improvements all bundled up in one new sensor, the OxyProbe II. The D540-VP has a straight VarioPin connector.The new membrane design is modeled after our successful 25 mm membrane cartridge. It has an expandable inner Silicone bladder that holds the electrolyte within a 316L stainless steel cage. During SIP processes or during autoclaving the electrolyte turns to steam, but does not distend the measurement membrane because the internal bladder expands first, taking the pressure off the sensitive measurement area. This allows us to have a maximum of electrolyte present, with minimum risk of damage to the sensor. The new membrane cartridge design is easy to use and durable enough to be used many times over if necessary.
Catalog No. NC0548926 Supplier Broadley James Corporation Supplier No. D540B120PTD9
After several years of refinement we are introducing our latest set of improvements all bundled up in one new sensor, the OxyProbe II. The D540-VP has a straight VarioPin connector.The new membrane design is modeled after our successful 25 mm membrane cartridge. It has an expandable inner Silicone bladder that holds the electrolyte within a 316L stainless steel cage. During SIP processes or during autoclaving the electrolyte turns to steam, but does not distend the measurement membrane because the internal bladder expands first, taking the pressure off the sensitive measurement area. This allows us to have a maximum of electrolyte present, with minimum risk of damage to the sensor. The new membrane cartridge design is easy to use and durable enough to be used many times over if necessary.