Learn More
Medchemexpress LLC Oxybenzone-13C6 (Benzophenone 3-13C6) | 131-57-7 | C813C6H12O3 | 1 MG
SDP
Catalog No. 5000433551
Encompass_Preferred
Oxybenzone-13C6 (Benzophenone 3-13C6) is the 13C-labeled Oxybenzone. It is a widely used UV filter in sun tans and skin protectants that acts as an endocrine-disrupting chemical (EDC) and can traverse the placental and blood-brain barriers. Benzophenone-3 has been shown to impair autophagy, alter epigenetic status, and disrupt retinoid X receptor signaling in apoptotic neuronal cells.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-33551 Supplier Medchemexpress LLC Supplier No. HYA0067S11MG
Oxybenzone-13C6 (Benzophenone 3-13C6) is the 13C-labeled Oxybenzone. It is a widely used UV filter in sun tans and skin protectants that acts as an endocrine-disrupting chemical (EDC) and can traverse the placental and blood-brain barriers. Benzophenone-3 has been shown to impair autophagy, alter epigenetic status, and disrupt retinoid X receptor signaling in apoptotic neuronal cells.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.