Learn More
Medchemexpress LLC Oxybenzone-d3 (benzophenone 3-d3) | 131-57-7 | MFCD00008387 | >95.0% | 231.26 g/mol | C14H9D3O3 | 2.5 MG
SDP
Catalog No. 5000284246
Encompass_Preferred
Oxybenzone-d3 (benzophenone 3-d3) is a deuterium-labeled stable isotope standard of oxybenzone intended for analytical and research use. Provided as a 2.5 mg solid, it serves as an internal standard for mass spectrometry, method development, and quantitation in environmental and cosmetic residue analysis.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Supplied as a 2.5 mg solid suitable for analytical use.
  • Chemical formula C14H9D3O3; molecular weight 231.26 g/mol.
  • Used for calibration, quantitation, and method validation.
  • Typically accompanied by vendor documentation such as a certificate of analysis.
Catalog No. 50-002-84246 Supplier Medchemexpress LLC Supplier No. HYW6539582.5MG
Oxybenzone-d3 (benzophenone 3-d3) is a deuterium-labeled stable isotope standard of oxybenzone intended for analytical and research use. Provided as a 2.5 mg solid, it serves as an internal standard for mass spectrometry, method development, and quantitation in environmental and cosmetic residue analysis.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Supplied as a 2.5 mg solid suitable for analytical use.
  • Chemical formula C14H9D3O3; molecular weight 231.26 g/mol.
  • Used for calibration, quantitation, and method validation.
  • Typically accompanied by vendor documentation such as a certificate of analysis.