Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxybenzone, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18604926 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-926 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-926 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B126125GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

B1261, 131-57-7, C14H12O3

Spectrum™ Chemical Oxybenzone, USP is one of the most widely used organic UVA filters in sunscreens. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
IUPAC Name 2-benzoyl-5-methoxyphenol
SMILES COC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1

Specifications

CAS Max % 1
Loss on Drying 0.02
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97.0 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.