Learn More
U.S. Pharmacopeia Oxybenzone, 131-57-7, MFCD00008387, 150mg
Supplier:  U.S. Pharmacopeia 1485001
Encompass
Molecular formula C14H12O3, Molecular weight 228.24, Melting Point 150.8 °F (66 °C) (congeals < 62 °C), Boiling Point 435.2 - 440.6 °F (224 - 227 °C), Flash point 212.00 °F (100.00 °C) Closed Cup, Synonyms: Benzophenone-3
Catalog No. NC9173214