Learn More
Apexbio Technology LLC Oxybuprocaine HCl 5987-82-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18055.1
Encompass_Preferred
Oxybuprocaine hydrochloride (CAS 5987-82-6) is a synthetic local anesthetic widely utilized in biomedical research focused on sensory neural pathways It acts by reversibly blocking voltage-gated sodium channels on neuronal membranes thereby inhibiting the initiation and transmission of nerve impulses This pharmacological action results in localized loss of sensation and is particularly advantageous in studies related to ocular and otolaryngological procedures Oxybuprocaine HCl serves as a valuable tool for investigating peripheral nerve function corneal responsiveness and models of nociception supporting research in neurophysiology and anesthetic pharmacology
Catalog No. 50-000-94705