Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxybuprocaine hydrochloride | 5987-82-6 | 99.9% | 1 ML
SDP

Catalog No. 5000411976
Encompass_Preferred

Oxybuprocaine hydrochloride (Benoxinate hydrochloride) reversibly blocks sodium channels and prevents propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is used especially in ophthalmology and otolaryngology.

  • Blocks sodium channels.
  • Prevents nerve impulse propagation in the cornea, conjunctiva, and sclera.
  • Used in ophthalmology and otolaryngology.
  • 10 mM, 1 mL solution in DMSO.
  • Purity of 99.93%.
  • Store at -80°C for 6 months or -20°C for 1 month in solvent, sealed, away from moisture.
  • Ship at room temperature if less than 2 weeks.

Catalog No. 50-004-11976 Supplier Medchemexpress LLC Supplier No. HYB128810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxybuprocaine hydrochloride (Benoxinate hydrochloride) reversibly blocks sodium channels and prevents propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is used especially in ophthalmology and otolaryngology.

  • Blocks sodium channels.
  • Prevents nerve impulse propagation in the cornea, conjunctiva, and sclera.
  • Used in ophthalmology and otolaryngology.
  • 10 mM, 1 mL solution in DMSO.
  • Purity of 99.93%.
  • Store at -80°C for 6 months or -20°C for 1 month in solvent, sealed, away from moisture.
  • Ship at room temperature if less than 2 weeks.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.