Learn More
Medchemexpress LLC Oxybuprocaine hydrochloride | 5987-82-6 | 99.9% | 250 MG
SDP
Catalog No. 5000414485
Encompass_Preferred
Oxybuprocaine hydrochloride, also known as Benoxinate hydrochloride, is a compound that reversibly blocks sodium channels. It prevents the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera, making it primarily used in ophthalmology and otolaryngology.
  • Reversibly blocks sodium channels
  • Prevents painful nerve impulses
  • Used in ophthalmology and otolaryngology
  • Soluble in DMSO and water
  • White to off-white solid appearance
Catalog No. 50-004-14485 Supplier Medchemexpress LLC Supplier No. 250MGHYB1288
Oxybuprocaine hydrochloride, also known as Benoxinate hydrochloride, is a compound that reversibly blocks sodium channels. It prevents the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera, making it primarily used in ophthalmology and otolaryngology.
  • Reversibly blocks sodium channels
  • Prevents painful nerve impulses
  • Used in ophthalmology and otolaryngology
  • Soluble in DMSO and water
  • White to off-white solid appearance