Learn More
Medchemexpress LLC Oxybuprocaine hydrochloride | 5987-82-6 | 99.9% | 50 MG
SDP
Catalog No. 5000411076
Encompass_Preferred
Oxybuprocaine hydrochloride is a sodium channel inhibitor that reversibly blocks sodium channels, preventing the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is primarily used in ophthalmology and otolaryngology.
  • Reversibly blocks sodium channels.
  • Prevents propagation of painful nerve impulses in the cornea, conjunctiva, and sclera.
  • Used in ophthalmology and otolaryngology.
  • High purity: 99.93%.
Catalog No. 50-004-11076 Supplier Medchemexpress LLC Supplier No. HYB128850MG
Oxybuprocaine hydrochloride is a sodium channel inhibitor that reversibly blocks sodium channels, preventing the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is primarily used in ophthalmology and otolaryngology.
  • Reversibly blocks sodium channels.
  • Prevents propagation of painful nerve impulses in the cornea, conjunctiva, and sclera.
  • Used in ophthalmology and otolaryngology.
  • High purity: 99.93%.