Learn More
Medchemexpress LLC Oxybuprocaine hydrochloride | 5987-82-6 | MFCD00012512 | 344.88 g/mol | C17H29ClN2O3 | 10 MG
SDP
Catalog No. 5000298146
Encompass_Preferred
Oxybuprocaine hydrochloride is a short-acting ester local anesthetic used topically to produce surface anesthesia by reversibly blocking voltage-gated sodium channels and preventing propagation of painful nerve impulses in ocular tissues. It is intended for research and analytical applications requiring a topical anesthetic agent.
  • Short-acting ester local anesthetic for topical applications.
  • Reversibly blocks voltage-gated sodium channels to prevent nerve impulse propagation.
  • Suitable for ophthalmic and otolaryngologic research applications.
  • HCl salt form with CAS 5987-82-6 and molecular weight 344.88 g/mol.
  • Supplied in a 10 MG research pack suitable for analytical and preclinical use.
Catalog No. 50-002-98146 Supplier Medchemexpress LLC Supplier No. HYB128810MG
Oxybuprocaine hydrochloride is a short-acting ester local anesthetic used topically to produce surface anesthesia by reversibly blocking voltage-gated sodium channels and preventing propagation of painful nerve impulses in ocular tissues. It is intended for research and analytical applications requiring a topical anesthetic agent.
  • Short-acting ester local anesthetic for topical applications.
  • Reversibly blocks voltage-gated sodium channels to prevent nerve impulse propagation.
  • Suitable for ophthalmic and otolaryngologic research applications.
  • HCl salt form with CAS 5987-82-6 and molecular weight 344.88 g/mol.
  • Supplied in a 10 MG research pack suitable for analytical and preclinical use.