Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxybuprocaine hydrochloride | 5987-82-6 | MFCD00012512 | 99.9% | 5 MG
SDP

Catalog No. 5000412510
Encompass_Preferred

Oxybuprocaine hydrochloride (Benoxinate hydrochloride) reversibly blocks sodium channels and prevents the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is especially used in ophthalmology and otolaryngology.

  • Reversibly blocks sodium channels.
  • Prevents propagation of painful nerve impulses.
  • Used in ophthalmology and otolaryngology.
  • Appearance: Solid, white to off-white.
  • Soluble in DMSO (100 mg/mL) and H2O (≥ 100 mg/mL).

Catalog No. 50-004-12510 Supplier Medchemexpress LLC Supplier No. HYB12885MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxybuprocaine hydrochloride (Benoxinate hydrochloride) reversibly blocks sodium channels and prevents the propagation of painful nerve impulses in the cornea, conjunctiva, and sclera. It is especially used in ophthalmology and otolaryngology.

  • Reversibly blocks sodium channels.
  • Prevents propagation of painful nerve impulses.
  • Used in ophthalmology and otolaryngology.
  • Appearance: Solid, white to off-white.
  • Soluble in DMSO (100 mg/mL) and H2O (≥ 100 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.