Learn More
Cayman Chemical Oxybuprocaine (hydrochloride), Purity: ≥95%, CAS: 5987-82-6, 25 g
SDP
Catalog No. 502942010
Encompass_Preferred
An anesthetic; reduces motor function, decreases righting behaviors in the placement test, and increases latency to paw withdrawal in the hot plate test when intrathecally administered in rats (ED50s = 0.43, 0.5, and 0.31 µmol/kg, respectively); reduces corneal sensitivity in the corneal touch test in dogs when topically applied at 0.4% w/v
Catalog No. 50-294-2010 Supplier Cayman Chemical Supplier No. 3997825 G
An anesthetic; reduces motor function, decreases righting behaviors in the placement test, and increases latency to paw withdrawal in the hot plate test when intrathecally administered in rats (ED50s = 0.43, 0.5, and 0.31 µmol/kg, respectively); reduces corneal sensitivity in the corneal touch test in dogs when topically applied at 0.4% w/v