Learn More
Medchemexpress LLC HY-B0267 500mg Medchemexpress, Oxybutynin CAS:5633-20-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0267500MG
Encompass_Preferred
Medchemexpress, HY-B0267 500mg Oxybutynin CAS:5633-20-5 Oxybutynin is an anticholinergic agent, which inhibits vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2435