Learn More
Apexbio Technology LLC Oxybutynin chloride 1508-65-2 100mg
SDP
Catalog No. 502240775
Encompass_Preferred
Oxybutynin chloride (CAS 1508-65-2) is a synthetic antimuscarinic agent that functions primarily by inhibiting the action of acetylcholine on muscarinic receptors in smooth muscle particularly within the bladder This antagonism results in reduced involuntary detrusor muscle contractions thereby modulating urinary urgency and frequency In biomedical research oxybutynin chloride is employed to investigate cholinergic pathways involved in bladder function neurogenic bladder disorders and the pharmacological modulation of muscarinic signaling in urinary physiology
This item is not returnable. View return policy
Catalog No. 50-224-0775 Supplier Apexbio Technology LLC Supplier No. B1134100
Oxybutynin chloride (CAS 1508-65-2) is a synthetic antimuscarinic agent that functions primarily by inhibiting the action of acetylcholine on muscarinic receptors in smooth muscle particularly within the bladder This antagonism results in reduced involuntary detrusor muscle contractions thereby modulating urinary urgency and frequency In biomedical research oxybutynin chloride is employed to investigate cholinergic pathways involved in bladder function neurogenic bladder disorders and the pharmacological modulation of muscarinic signaling in urinary physiology