Learn More
Medchemexpress LLC HY-B0267A 100mg Medchemexpress, Oxybutynin (chloride) CAS:1508-65-2 Purity:>98%
SDP
Catalog No. 502082436
Encompass_Preferred
Medchemexpress, HY-B0267A 100mg Oxybutynin (chloride) CAS:1508-65-2 Oxybutynin chloride is an anticholinergic agent, which inhibits vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2436 Supplier Medchemexpress LLC Supplier No. HYB0267A100MG
Medchemexpress, HY-B0267A 100mg Oxybutynin (chloride) CAS:1508-65-2 Oxybutynin chloride is an anticholinergic agent, which inhibits vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.