Learn More
Medchemexpress LLC Oxybutynin-d11 chloride | 1185151-95-4 | 97.4% | 1 MG
Supplier: Medchemexpress LLC HYB0267AS1MG
Oxybutynin-d11 (chloride) is a deuterium-labeled version of Oxybutynin chloride. Oxybutynin chloride is an anticholinergic agent known to inhibit vascular Kv channels in a concentration-dependent manner, with an IC50 of 11.51 μM. This compound is intended for research use only and is not sold to patients.
- Can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
- Stable heavy isotopes have been incorporated into drug molecules for use as tracers for quantitation during drug development.
- Deuteration can affect pharmacokinetic and metabolic profiles of drugs.