Learn More
Medchemexpress LLC Oxybutynin-d11 chloride | 1185151-95-4 | MFCD06658724 | 97.4% | 405.02 g·mol⁻¹ | C22H21D11ClNO3 | 10 MG
SDP
Catalog No. 5000297074
Encompass_Preferred
Oxybutynin-d11 chloride is a deuterium-labeled isotopologue of oxybutynin chloride intended as a stable-isotope reference standard for analytical, pharmacokinetic, and mass-spectrometry studies. Supplied as the chloride salt with defined molecular weight and high purity, it is suitable for use as an internal standard to improve quantitation and identification in LC-MS and GC-MS workflows.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • High chemical purity (97.4%) and isotopic labeling for reliable quantitation.
  • Supplied as a chloride salt with known molecular formula and molecular weight for accurate calculations.
  • Soluble in DMSO for convenient sample preparation.
Catalog No. 50-002-97074 Supplier Medchemexpress LLC Supplier No. HYB0267AS10MG
Add to Cart
Add to Cart
Oxybutynin-d11 chloride is a deuterium-labeled isotopologue of oxybutynin chloride intended as a stable-isotope reference standard for analytical, pharmacokinetic, and mass-spectrometry studies. Supplied as the chloride salt with defined molecular weight and high purity, it is suitable for use as an internal standard to improve quantitation and identification in LC-MS and GC-MS workflows.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • High chemical purity (97.4%) and isotopic labeling for reliable quantitation.
  • Supplied as a chloride salt with known molecular formula and molecular weight for accurate calculations.
  • Soluble in DMSO for convenient sample preparation.