Learn More
Cayman Chemical Oxybutynin-d10 (hydrochloride), Purity: ≥99% deuterated forms (d1-d10), CAS: 120092-65-1, 1 mg
SDP
Catalog No. 502890104
Encompass_Preferred
An internal standard for the quantification of oxybutynin by GC- or LC-MS
Catalog No. 50-289-0104 Supplier Cayman Chemical Supplier No. 100106551 MG
An internal standard for the quantification of oxybutynin by GC- or LC-MS