Learn More
Cayman Chemical Oxybutynin (hydrochloride), Purity: ≥98%, CAS: 1508-65-2, 1 g
SDP
Catalog No. 502994808
Encompass_Preferred
An antagonist of muscarinic acetylcholine receptors (Kis = 5, 14.5, 3.7, 5.3, and 40 nM for human recombinant M1-5, respectively); inhibits carbachol- induced intracellular calcium mobilization in bladder smooth muscle and submandibular gland cells isolated from cynomolgus monkeys (Kis = 2 and 1 nM, respectively); inhibits VIBC and OIS in rats (ID50s = 0.062 and 0.089 mg/kg, respectively); increases PD and LMA (ED50s = 0.29 and 0.52 mg/kg, respectively) and decreases SIT (ID50 = 0.22 mg/kg) in rats
Catalog No. 50-299-4808 Supplier Cayman Chemical Supplier No. 238251 G
An antagonist of muscarinic acetylcholine receptors (Kis = 5, 14.5, 3.7, 5.3, and 40 nM for human recombinant M1-5, respectively); inhibits carbachol- induced intracellular calcium mobilization in bladder smooth muscle and submandibular gland cells isolated from cynomolgus monkeys (Kis = 2 and 1 nM, respectively); inhibits VIBC and OIS in rats (ID50s = 0.062 and 0.089 mg/kg, respectively); increases PD and LMA (ED50s = 0.29 and 0.52 mg/kg, respectively) and decreases SIT (ID50 = 0.22 mg/kg) in rats