Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Oxycodone Control

Catalog No. 01801158 Shop All Beckman Coulter Products
Change view
Click to view available options
Concentration:
100 ng/mL
300 ng/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
01-801-158 300 ng/mL
01-801-157 100 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-801-158 Supplier Beckman Coulter™ Supplier No. A87935
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxycodone Control for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

Concentration 300 ng/mL
For Use With (Equipment) Beckman Coulter Analyzers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.