Learn More
AdipoGen Oxydemeton-methyl
SDP
Supplier:  AdipoGen CDXO0187M050
Encompass_Preferred
Chemical. CAS 301-12-2. Formula C6H15O4PS2. MW 246.28. Oxydemeton-methyl is an organothiophosphate insecticide. Acetylcholinesterase AChE inhibitor in the nervous system. Primarily used to control aphids, mites, and thrips. Compound can be used as analytical reference material.
Catalog No. 50-256-0294