Learn More
Cambridge Isotope Laboratories Oxydemeton-methyl (di-o-methyl-D6 98%) 100 ug/mL in acetonitrile 1 2 mL
Catalog No. 5000166908
Encompass
Oxydemeton-methyl (di-o-methyl-D6 98%) 100 ug/mL in acetonitrile 1 2 mL
Catalog No. 50-001-66908 Supplier Cambridge Isotope Laboratories Supplier No. DLM71501.2
Oxydemeton-methyl (di-o-methyl-D6 98%) 100 ug/mL in acetonitrile 1 2 mL