Learn More
Medchemexpress LLC Oxyfedrine hydrochloride | 16777-42-7 | 99.9% | 349.85 | C19H24ClNO3 | 10 MG
SDP
Catalog No. 5000243979
Encompass_Preferred
Oxyfedrine hydrochloride is a vasodilator and an orally active β-adrenoreceptor agonist commonly used in research to investigate cardiovascular function and adrenergic signaling. The compound is supplied as a high-purity hydrochloride salt in small-scale research quantities suitable for biochemical and pharmacological studies.
  • High purity suitable for research use.
  • Orally active β-adrenoreceptor agonist.
  • Available in multiple small-scale package sizes, including 10 mg.
  • Stable storage at 4°C in a sealed container away from moisture and light.
  • Recommended solvent storage: -80°C for long-term, -20°C for short-term.
  • Molecular formula C19H24ClNO3; molecular weight 349.85.
Catalog No. 50-002-43979 Supplier Medchemexpress LLC Supplier No. HY10830010MG
Oxyfedrine hydrochloride is a vasodilator and an orally active β-adrenoreceptor agonist commonly used in research to investigate cardiovascular function and adrenergic signaling. The compound is supplied as a high-purity hydrochloride salt in small-scale research quantities suitable for biochemical and pharmacological studies.
  • High purity suitable for research use.
  • Orally active β-adrenoreceptor agonist.
  • Available in multiple small-scale package sizes, including 10 mg.
  • Stable storage at 4°C in a sealed container away from moisture and light.
  • Recommended solvent storage: -80°C for long-term, -20°C for short-term.
  • Molecular formula C19H24ClNO3; molecular weight 349.85.