Learn More
Medchemexpress LLC Oxyfedrine hydrochloride | 16777-42-7 | 99.9% | 349.85 | C19H24ClNO3 | 25 MG
SDP
Catalog No. 5000242483
Encompass_Preferred
Oxyfedrine hydrochloride is the hydrochloride salt of oxyfedrine, an orally active beta-adrenoreceptor agonist and vasodilator used in pharmacological research. Supplied as a solid research-grade powder, it includes batch analytical data to support experimental use and handling.
  • High purity powder (~99.9%).
  • Acts as a beta-adrenoreceptor agonist and vasodilator for pharmacology studies.
  • Solid form with defined molecular weight and formula for dosing calculations.
  • Stable when stored sealed, protected from moisture and light.
  • Provided with batch-specific analytical documents (COA, HNMR, LCMS).
Catalog No. 50-002-42483 Supplier Medchemexpress LLC Supplier No. HY10830025MG
Oxyfedrine hydrochloride is the hydrochloride salt of oxyfedrine, an orally active beta-adrenoreceptor agonist and vasodilator used in pharmacological research. Supplied as a solid research-grade powder, it includes batch analytical data to support experimental use and handling.
  • High purity powder (~99.9%).
  • Acts as a beta-adrenoreceptor agonist and vasodilator for pharmacology studies.
  • Solid form with defined molecular weight and formula for dosing calculations.
  • Stable when stored sealed, protected from moisture and light.
  • Provided with batch-specific analytical documents (COA, HNMR, LCMS).