Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyfedrine hydrochloride | 16777-42-7 | 99.9% | 349.85 | C19H24ClNO3 | 50 MG
SDP

Catalog No. 5000244079
Encompass_Preferred

Oxyfedrine hydrochloride is an orally active β-adrenoreceptor agonist and coronary vasodilator supplied as the hydrochloride salt for research use. It is intended for pharmacological and cardiovascular studies and should be stored and handled according to supplier recommendations.

  • High purity for research applications (99.9% as listed).
  • Molecular formula C19H24ClNO3 and molecular weight 349.85.
  • Supplied as a solid, facilitating handling and formulation.
  • Storage: sealed, away from moisture and light; in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for studies of β-adrenergic activity and coronary vasodilation.

Catalog No. 50-002-44079 Supplier Medchemexpress LLC Supplier No. HY10830050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyfedrine hydrochloride is an orally active β-adrenoreceptor agonist and coronary vasodilator supplied as the hydrochloride salt for research use. It is intended for pharmacological and cardiovascular studies and should be stored and handled according to supplier recommendations.

  • High purity for research applications (99.9% as listed).
  • Molecular formula C19H24ClNO3 and molecular weight 349.85.
  • Supplied as a solid, facilitating handling and formulation.
  • Storage: sealed, away from moisture and light; in solvent -80°C (6 months) or -20°C (1 month).
  • Suitable for studies of β-adrenergic activity and coronary vasodilation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.