Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxyfedrine hydrochloride | 16777-42-7 | MFCD01695148 | 99.9% | 349.85 | C19H24ClNO3 | 100 MG
SDP

Catalog No. 5000242494
Encompass_Preferred

Oxyfedrine hydrochloride is the hydrochloride salt of oxyfedrine, a vasodilator and orally active β-adrenoreceptor agonist supplied for research use. It is intended for pharmacology and biochemical studies of adrenergic activity and vascular effects, with available documentation to support analytical and safety review.

  • CAS number 16777-42-7.
  • Chemical formula C19H24ClNO3.
  • Molecular weight 349.85.
  • High purity (99.9% by HPLC).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Includes COA and SDS documentation.
  • Suitable for pharmacology and adrenergic receptor research.

Catalog No. 50-002-42494 Supplier Medchemexpress LLC Supplier No. HY108300100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyfedrine hydrochloride is the hydrochloride salt of oxyfedrine, a vasodilator and orally active β-adrenoreceptor agonist supplied for research use. It is intended for pharmacology and biochemical studies of adrenergic activity and vascular effects, with available documentation to support analytical and safety review.

  • CAS number 16777-42-7.
  • Chemical formula C19H24ClNO3.
  • Molecular weight 349.85.
  • High purity (99.9% by HPLC).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Includes COA and SDS documentation.
  • Suitable for pharmacology and adrenergic receptor research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.