Learn More
Bellco Glass, Inc. OXYFERM DO Probe,12x425mm Hamilton
SDP
Catalog No. 501946410
Encompass_Preferred
Dissolved oxygen DO and PH electrodes are the eyes and ears for many biotechnology process high quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world.Bellco offers Hamilton Gel-Filled pH and stainless steel polarographic dissolved oxygen sensors withthe standard 4 pin, T82, K8 and VP style connectors in 120 to 425 mm lengths.
Catalog No. 50-194-6410 Supplier Bellco Glass, Inc. Supplier No. 780831425
Add to Cart
Add to Cart
Dissolved oxygen DO and PH electrodes are the eyes and ears for many biotechnology process high quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world.Bellco offers Hamilton Gel-Filled pH and stainless steel polarographic dissolved oxygen sensors withthe standard 4 pin, T82, K8 and VP style connectors in 120 to 425 mm lengths.