Learn More
Bellco Glass, Inc. OXYFERM VP DO Probe,12x225mm With VP Connector - Hamilton
SDP
Catalog No. 501946413
Encompass_Preferred
Dissolved oxygen DO and PH electrodes are the eyes and ears for many biotechnology process high quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world.Bellco offers Hamilton Gel-Filled pH and stainless steel polarographic dissolved oxygen sensors withthe standard 4 pin, T82, K8 and VP style connectors in 120 to 425 mm lengths.
Catalog No. 50-194-6413 Supplier Bellco Glass, Inc. Supplier No. 780832225
Dissolved oxygen DO and PH electrodes are the eyes and ears for many biotechnology process high quality sensors are required for critical measurement and Hamilton sensors are some of the fi in the world.Bellco offers Hamilton Gel-Filled pH and stainless steel polarographic dissolved oxygen sensors withthe standard 4 pin, T82, K8 and VP style connectors in 120 to 425 mm lengths.