Learn More
Oxyrase Oxyfluor 5Ml Bottle 1/Pk
SDP
Catalog No. 503002016
Encompass
Formulated specifically for fluorescence microscopy. Prevents photo-bleaching and photo-damage by removing dissolved oxygen. Significantly reduces/eliminates formation of oxygen free radicals. Results in brighter cell images. Provides protective effect for biological material being studied. Excellent solution for live cell imaging and cell culture applications.
This item is not returnable. View return policy
Catalog No. 50-300-2016 Supplier Oxyrase Supplier No. OF0005
Formulated specifically for fluorescence microscopy. Prevents photo-bleaching and photo-damage by removing dissolved oxygen. Significantly reduces/eliminates formation of oxygen free radicals. Results in brighter cell images. Provides protective effect for biological material being studied. Excellent solution for live cell imaging and cell culture applications.