Learn More
Medchemexpress LLC Oxyfluorfen | 42874-03-3 | 99.7 % | 100 MG
SDP
Catalog No. 5000233170
Encompass_Preferred
Oxyfluorfen is a diphenyl ether herbicide and protoporphyrinogen oxidase (PPO) inhibitor supplied as a research-grade solid for laboratory studies of herbicidal activity, environmental fate, and toxicology.
  • High purity suitable for analytical and research use.
  • Off-white to light yellow solid physical form.
  • Available in small pack sizes for laboratory experiments.
  • Recommended cold storage to maintain stability.
  • Acts as a PPO inhibitor affecting chlorophyll synthesis for mechanism studies.
Catalog No. 50-002-33170 Supplier Medchemexpress LLC Supplier No. HY119176100MG
Oxyfluorfen is a diphenyl ether herbicide and protoporphyrinogen oxidase (PPO) inhibitor supplied as a research-grade solid for laboratory studies of herbicidal activity, environmental fate, and toxicology.
  • High purity suitable for analytical and research use.
  • Off-white to light yellow solid physical form.
  • Available in small pack sizes for laboratory experiments.
  • Recommended cold storage to maintain stability.
  • Acts as a PPO inhibitor affecting chlorophyll synthesis for mechanism studies.