Learn More
Medchemexpress LLC Oxyfluorfen | 42874-03-3 | 99.7% | 50 MG
SDP
Catalog No. 5000231893
Encompass_Preferred
Oxyfluorfen is a diphenyl ether herbicide supplied as a research-grade analytical reagent. It inhibits protoporphyrinogen oxidase (PPO), disrupting chlorophyll synthesis, and is used in analytical, environmental, and toxicology studies.
  • High purity analytical grade (99.7%).
  • Molecular formula C15H11ClF3NO4; molecular weight 361.70 g/mol.
  • Supplied in small quantities suitable for laboratory testing.
  • Used to study PPO inhibition, herbicidal activity, and environmental fate.
  • Reported to induce DNA damage and is toxic to aquatic organisms; handle with appropriate precautions.
Catalog No. 50-002-31893 Supplier Medchemexpress LLC Supplier No. HY11917650MG
Oxyfluorfen is a diphenyl ether herbicide supplied as a research-grade analytical reagent. It inhibits protoporphyrinogen oxidase (PPO), disrupting chlorophyll synthesis, and is used in analytical, environmental, and toxicology studies.
  • High purity analytical grade (99.7%).
  • Molecular formula C15H11ClF3NO4; molecular weight 361.70 g/mol.
  • Supplied in small quantities suitable for laboratory testing.
  • Used to study PPO inhibition, herbicidal activity, and environmental fate.
  • Reported to induce DNA damage and is toxic to aquatic organisms; handle with appropriate precautions.