Learn More
Medchemexpress LLC Oxyfluorfen | 42874-03-3 | 99.7% | 500 MG
SDP
Catalog No. 5000233456
Encompass_Preferred
Oxyfluorfen is a diphenyl ether herbicide supplied as a 500 mg research-grade reagent. It functions as a protoporphyrinogen oxidase (PPO) inhibitor and is intended for laboratory research use only; not for human or therapeutic applications.
  • High purity (99.7%) suitable for analytical and research use.
  • Supplied as a 500 mg vial for small-scale studies.
  • Chemical formula C15H11ClF3NO4; molecular weight 361.70 g/mol.
  • Acts as a PPO inhibitor useful in plant physiology and mechanism studies.
  • For research use only; not for human consumption or therapeutic use.
Catalog No. 50-002-33456 Supplier Medchemexpress LLC Supplier No. HY119176500MG
Oxyfluorfen is a diphenyl ether herbicide supplied as a 500 mg research-grade reagent. It functions as a protoporphyrinogen oxidase (PPO) inhibitor and is intended for laboratory research use only; not for human or therapeutic applications.
  • High purity (99.7%) suitable for analytical and research use.
  • Supplied as a 500 mg vial for small-scale studies.
  • Chemical formula C15H11ClF3NO4; molecular weight 361.70 g/mol.
  • Acts as a PPO inhibitor useful in plant physiology and mechanism studies.
  • For research use only; not for human consumption or therapeutic use.