Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill
SDP

Catalog No. 23254447
Change view
Click to view available options
Quantity:
30 Ampules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-254447 30 Ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-254447 Supplier CHEMetrics Supplier No. R7518
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides results in two minutes or less

  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life

Specifications

Analytes Dissolved Oxygen (DO)
Format Liquid
For Use With (Equipment) C-7518 comparators
No. of Tests 30
Quantity 30 Ampules
Range 5 to 180 ppb
Test Method Rhodazine D
Test Time 2 min.
Type CHEMets Dissolved Oxygen Refill
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Warning! Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.