Learn More
Perkin Elmer US LLC Oxygen Dual-Stage High-Purity Regulator
Supplier:  Perkin Elmer US LLC N8152491
Encompass_Preferred
Benefits and Features: High-purity nickel plated brass barstock body High-purity stainless steel diaphragm Metal to metal seals minimize diffusion of contaminants 2-1/2 inlet and delivery pressure gauges Equipped with outlet diaphragm valve Bonnet ported and threaded to pipe gases away from work area 1/4 compression tube outlet connection 10 micron metal filter protects seat from contamination Panel mountable, front bonnet and back bonnet threaded Bonnet vent holes size: 1/8 FNPT
Catalog No. 50-210-3933