Learn More
Euthanex Oxygen Hose, Green
Catalog No. 501540159
Encompass_Preferred
Oxygen Hose, Green. Standard 6-foot hose connection from oxygen regulator to the anesthesia system. Green hose is fitted with female DISS fittings on both ends.
Catalog No. 50-154-0159 Supplier Euthanex Supplier No. EZ15021
Oxygen Hose, Green. Standard 6-foot hose connection from oxygen regulator to the anesthesia system. Green hose is fitted with female DISS fittings on both ends.