Learn More
Euthanex Oxygen Output Splitter
Catalog No. 501540158
Encompass_Preferred
Oxygen Output Splitter is a "Y-splitter" for oxygen regulator output or wall outputs with DISS connection. This splitter allows two Anesthesia systems to connect to a single oxygen source. An on-demand check valve in both outputs allows the use of a single system when required.
Catalog No. 50-154-0158 Supplier Euthanex Supplier No. EZ124DV
Oxygen Output Splitter is a "Y-splitter" for oxygen regulator output or wall outputs with DISS connection. This splitter allows two Anesthesia systems to connect to a single oxygen source. An on-demand check valve in both outputs allows the use of a single system when required.