Learn More
Euthanex EZ-320 Oxygen Regulator with H-Cylinder fitting, 50 psi, for H-Cylinder O2 Tanks using a CGA-540 threaded connection.
Catalog No. NC1164342
Encompass_Preferred
EZ-320 Oxygen Regulator with H-Cylinder fitting is factory preset to deliver 50 psi and attaches to large oxygen tanks ("H-Cylinders") using a CGA-540 threaded connection.
Catalog No. NC1164342 Supplier Euthanex Supplier No. EZ320
EZ-320 Oxygen Regulator with H-Cylinder fitting is factory preset to deliver 50 psi and attaches to large oxygen tanks ("H-Cylinders") using a CGA-540 threaded connection.