Learn More
METTLER TOLEDO DO Sensor InPro6860i/12/220/nA
Catalog No. NC0391051 Shop All METTLER TOLEDO Products
Encompass
The InPro6860i is an optical dissolved oxygen sensor which offers highly accurate oxygen measurements with enhanced stability, suitable for biopharma applications.
Catalog No. NC0391051 Supplier METTLER TOLEDO Supplier No. 30014101
The InPro6860i is an optical dissolved oxygen sensor which offers highly accurate oxygen measurements with enhanced stability, suitable for biopharma applications.