Learn More
World Precision Instrument Oxygen Sensor-FLOX-PATCH
SDP
Catalog No. 502538806
Encompass
FLOX is a device for measuring fluorescence lifetime, phase and intensity. It uses LED excitation and photodiode detection with filter-based wavelength selection for easy experimental set-up and control.
FLOX is especially useful for oxygen sensing applications where stability and sensitivity to drift is important and where sample set-ups must be left undisturbed for long periods of time.
Catalog No. 50-253-8806 Supplier World Precision Instrument Supplier No. FLOXPATCH
FLOX is a device for measuring fluorescence lifetime, phase and intensity. It uses LED excitation and photodiode detection with filter-based wavelength selection for easy experimental set-up and control.
FLOX is especially useful for oxygen sensing applications where stability and sensitivity to drift is important and where sample set-ups must be left undisturbed for long periods of time.