Learn More
Med Vet International Smart Oxygen™ Portable ICU, J1509, Each
Catalog No. 5000495925
Encompass_Preferred
Features: • Lightweight metal cage is completely covered with clear vinyl canopy • Various portals allow administration of oxygen • Nebulization equipment also included • Plastic tray covers bottom of unit • Opens on top and zippered close for tight seal • 25lb patient Capacity Suggested Use • Respiratory distress • Pre and post anesthesia • Nebulization for treatment of respiratory disease
Catalog No. 50-004-95925 Supplier Med Vet International Supplier No. JOR1509
Features: • Lightweight metal cage is completely covered with clear vinyl canopy • Various portals allow administration of oxygen • Nebulization equipment also included • Plastic tray covers bottom of unit • Opens on top and zippered close for tight seal • 25lb patient Capacity Suggested Use • Respiratory distress • Pre and post anesthesia • Nebulization for treatment of respiratory disease