Learn More
Apexbio Technology LLC Oxymatrine 16837-52-8 1g
SDP
Catalog No. 5000090717
Encompass_Preferred
Oxymatrine (CAS 16837-52-8) is a quinolizidine alkaloid extracted from Sophora flavescens It has been shown to modulate multiple cellular pathways associated with fibrosis and neurodegeneration including inhibition of TGF- 1-mediated signaling and reduction of pro-inflammatory cytokine expression Oxymatrine exhibits antifibrotic and neuroprotective properties in various experimental models It is also extensively studied for its antiviral activity against hepatitis B virus and has been applied in traditional Chinese medicine for hepatitis management This compound is valuable for investigating mechanisms of fibrosis neuroprotection and viral pathogenesis
Catalog No. 50-000-90717 Supplier Apexbio Technology LLC Supplier No. N18351000
Oxymatrine (CAS 16837-52-8) is a quinolizidine alkaloid extracted from Sophora flavescens It has been shown to modulate multiple cellular pathways associated with fibrosis and neurodegeneration including inhibition of TGF- 1-mediated signaling and reduction of pro-inflammatory cytokine expression Oxymatrine exhibits antifibrotic and neuroprotective properties in various experimental models It is also extensively studied for its antiviral activity against hepatitis B virus and has been applied in traditional Chinese medicine for hepatitis management This compound is valuable for investigating mechanisms of fibrosis neuroprotection and viral pathogenesis