Learn More
Selleck Chemical LLC Oxymatrine 100mg 16837-52-8 Matrine N-oxide
SDP
Catalog No. 505506
Encompass_Preferred
Oxymatrine (Matrine N-oxide) is a quinolizidine alkaloid extracted from the root of Sophora flavescens with anti-inflammatory, antifibrosis, and antitumor effects, inhibits the iNOS expression and TGF-?/Smad pathway. It is used for the treatment of viral hepatitis, cancer, viral myocarditis, gastrointestinal hemorrhage and skin diseases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-550-6 Supplier Selleck Chemical LLC Supplier No. S2338100MG
Oxymatrine (Matrine N-oxide) is a quinolizidine alkaloid extracted from the root of Sophora flavescens with anti-inflammatory, antifibrosis, and antitumor effects, inhibits the iNOS expression and TGF-?/Smad pathway. It is used for the treatment of viral hepatitis, cancer, viral myocarditis, gastrointestinal hemorrhage and skin diseases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.