Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxypeucedanin | 737-52-0 | 99.5% | 286.28 | C16H14O5 | 10 MG
SDP

Catalog No. 5000295877
Encompass_Preferred

Oxypeucedanin is a natural furanocoumarin research compound reported to inhibit hKv1.5 channel currents and modulate PI3K/AKT/NF-κB and MAPK pathways. It is supplied as a high-purity solid for in vitro research; documentation includes a datasheet, certificate of analysis, and safety data sheet.

  • Natural furanocoumarin research compound.
  • CAS number 737-52-0; molecular weight 286.28 g/mol.
  • Reported purity 99.5% (HPLC).
  • Supplied as a 10 MG quantity suitable for in vitro studies.
  • Documentation available: datasheet, COA, SDS.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-95877 Supplier Medchemexpress LLC Supplier No. HYN074710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxypeucedanin is a natural furanocoumarin research compound reported to inhibit hKv1.5 channel currents and modulate PI3K/AKT/NF-κB and MAPK pathways. It is supplied as a high-purity solid for in vitro research; documentation includes a datasheet, certificate of analysis, and safety data sheet.

  • Natural furanocoumarin research compound.
  • CAS number 737-52-0; molecular weight 286.28 g/mol.
  • Reported purity 99.5% (HPLC).
  • Supplied as a 10 MG quantity suitable for in vitro studies.
  • Documentation available: datasheet, COA, SDS.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.