Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxypeucedanin hydrate | 2643-85-8 | 5 MG
SDP

Catalog No. 5000420894
Encompass_Preferred

Oxypeucedanin hydrate is a natural product isolated from D. anethifolia. It exhibits mild toxicity on fibroblasts and parental lymphoma cells. This product is for research use only.

  • Isolated from D. anethifolia
  • Exhibits mild toxicity on fibroblasts and parental lymphoma cells
  • Molecular weight of 304.29
  • Chemical formula is C16H16O6
  • Appears as an off-white to light yellow solid
  • Purity of 98.0%
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-20894 Supplier Medchemexpress LLC Supplier No. HYN26225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxypeucedanin hydrate is a natural product isolated from D. anethifolia. It exhibits mild toxicity on fibroblasts and parental lymphoma cells. This product is for research use only.

  • Isolated from D. anethifolia
  • Exhibits mild toxicity on fibroblasts and parental lymphoma cells
  • Molecular weight of 304.29
  • Chemical formula is C16H16O6
  • Appears as an off-white to light yellow solid
  • Purity of 98.0%
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.