Learn More
Medchemexpress LLC (-)-Oxypeucedanin hydrate ((-)-Prangol) | 133164-11-1 | 5 MG
SDP
Catalog No. 5000421235
Encompass_Preferred
(-)-Oxypeucedanin hydrate, also known as (-)-Prangol, is a furocoumarin derivative isolated from Ducrosia anethifolia. This compound demonstrates significant antiproliferative and cytotoxic activities, making it suitable for various research applications.
  • Furocoumarin derivative
  • Isolated from Ducrosia anethifolia
  • Exhibits antiproliferative activity
  • Exhibits cytotoxic activity
Catalog No. 50-004-21235 Supplier Medchemexpress LLC Supplier No. HYN22545MG
(-)-Oxypeucedanin hydrate, also known as (-)-Prangol, is a furocoumarin derivative isolated from Ducrosia anethifolia. This compound demonstrates significant antiproliferative and cytotoxic activities, making it suitable for various research applications.
  • Furocoumarin derivative
  • Isolated from Ducrosia anethifolia
  • Exhibits antiproliferative activity
  • Exhibits cytotoxic activity