Learn More
Cayman Chemical (+)-Oxypeucedanin hydrate, Purity: ≥98%, CAS: 2643-85-8, 50 mg
SDP
Catalog No. 502950148
Encompass_Preferred
A furanocoumarin; active against a variety of bacteria and fungi (MICs = 9.76-625 µg/ml); inhibits proliferation of human MK-1 gastric and HeLa cervical cancer cells, as well as murine B16/F10 melanoma cells (EC50s = 47.2, 80.3, and 42 µg/ml, respectively); also inhibits proliferation of sensitive and multidrug-resistant L5178Y murine lymphoma cells (IC50s = 41.96 and 60.58 µM, respectively)
Catalog No. 50-295-0148 Supplier Cayman Chemical Supplier No. 3384150 MG
A furanocoumarin; active against a variety of bacteria and fungi (MICs = 9.76-625 µg/ml); inhibits proliferation of human MK-1 gastric and HeLa cervical cancer cells, as well as murine B16/F10 melanoma cells (EC50s = 47.2, 80.3, and 42 µg/ml, respectively); also inhibits proliferation of sensitive and multidrug-resistant L5178Y murine lymphoma cells (IC50s = 41.96 and 60.58 µM, respectively)