Learn More
Medchemexpress LLC Oxyphenbutazone monohydrate | 7081-38-1 | 98.0% | 50 MG
SDP
Catalog No. 5000412937
Encompass_Preferred
Oxyphenbutazone monohydrate is a metabolite with anti-inflammatory effects. It acts as an orally active non-selective COX inhibitor and can selectively eliminate non-replicating Mycobacterium tuberculosis.
  • Anti-inflammatory effect.
  • Orally active.
  • Non-selective COX inhibitor.
  • Selectively kills non-replicating Mycobacterium tuberculosis.
  • Enhances anticancer efficiency in certain cells.
  • Exhibits reparative effects in hepatocytes.
Catalog No. 50-004-12937 Supplier Medchemexpress LLC Supplier No. HYB135550MG
Oxyphenbutazone monohydrate is a metabolite with anti-inflammatory effects. It acts as an orally active non-selective COX inhibitor and can selectively eliminate non-replicating Mycobacterium tuberculosis.
  • Anti-inflammatory effect.
  • Orally active.
  • Non-selective COX inhibitor.
  • Selectively kills non-replicating Mycobacterium tuberculosis.
  • Enhances anticancer efficiency in certain cells.
  • Exhibits reparative effects in hepatocytes.